Learn More
Lamin B1 Mouse anti-Human, CoraLite 488, Clone: 3C10G12, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL488-66095-100UL
Descrizione
The Lamin B1 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human Lamin B1. This antibody recognizes human, mouse, rat antigen. The Lamin B1 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specifica
| Lamin B1 | |
| Monoclonal | |
| 2000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| 3C10G12 | |
| CoraLite 488 | |
| 100 μL | |
| 4001 | |
| IgG1 |