missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Laminin beta-1 Recombinant Rabbit Monoclonal Antibody (6I9T4)

Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen™ MA535495
Questo articolo non è restituibile.
Consulta la politica di reso
Beschreibung
Immunogen sequence: KKTLDGELDE KYKKVENLIA KKTEESADAR RKAEMLQNEA KTLLAQANSK LQLLKDLERK YEDNQRYLED KAQELARLEG EVRSLLKDIS QKVAVYSTCL.
Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.
Spezifikation
| Laminin beta-1 | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P02469, P07942 | |
| LAMB1 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| 6I9T4 | |
| Unconjugated | |
| LAMB1 | |
| C77966; C80098; C81607; CLM; D130003D08Rik; I79_017505; LAMB1; Lamb-1; LAMB1; basement membrane protein; Lamb1-1; laminin B1; laminin B1 chain; laminin B1 subunit 1; laminin beta 1; laminin beta-1 chain; laminin subunit beta 1; laminin subunit beta-1; laminin, beta 1; laminin-1 subunit beta; Laminin-10 subunit beta; laminin-12 subunit beta; Laminin-2 subunit beta; laminin-6 subunit beta; Laminin-8 subunit beta; LIS5; LOW QUALITY PROTEIN: laminin subunit beta-1; MGC142015 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 16777, 298941, 3912 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur