Learn More
LAP (TGF-beta 1) Antibody (1015022), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals FAB77541A
Codice prodotto. 18629803
  • 693.00€ / 0.10mL
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
LAP (TGF-beta 1) Monoclonal antibody specifically detects LAP (TGF-beta 1) in Human samples. It is validated for ELISA, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifica
Specifications
LAP (TGF-beta 1)
Monoclonal
APC
PBS
Mouse
Protein A or G purified from cell culture supernatant
RUO
Primary
Human
Purified
ELISA, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
1015022
ELISA, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1
Chinese hamster ovary cell line CHO-derived recombinant human LAP (TGF-beta 1), Met1-Ser390, Accession # P01137
0.1 mL
Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Signal Transduction
7040
Store at 4°C in the dark.
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.