Learn More
LABPLAS Large Rack
Codice prodotto. 17930205
Click to view available options
Include:
None
Sterile Bags
Dimensione della confezione:
Pezzo
pezzi 3
Codice prodotto. 17930205
Marca: LABPLAS SA57012
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Large rack designed for the safe transport or storage of sample bags.
  • Facilitates the transport and maintenance of sample bags
  • Foldable, lightweight, sturdy, easy to store and dishwasher safe
  • Ideal for sampling activities in the laboratory or in the field
  • Allows you to keep your hands free as you work with sample bags
Specifica
Include None
Product Type Bag Rack
Autoclavabile Not Autoclavable
Capacità Bags