Learn More
LABPLAS Large Rack
Click to view available options
Include:
None
Sterile Bags
Dimensione della confezione:
Pezzo
pezzi 3
Descrizione
- Facilitates the transport and maintenance of sample bags
- Foldable, lightweight, sturdy, easy to store and dishwasher safe
- Ideal for sampling activities in the laboratory or in the field
- Allows you to keep your hands free as you work with sample bags
Specifica
Specifica
| Include | None |
| Product Type | Bag Rack |
| Autoclavabile | Not Autoclavable |
| Capacità | Bags |