Learn More
Gilac™ Large Volume Container with Pivoting Wheels and Brakes
Click to view available options
Capacità (sistema metrico):
100 L
170 L
220 L
310 L
500 L
Include:
2 Pivoting Wheels with Brakes
4 Pivoting Wheels, 2 Brakes
Dimensione della confezione:
Pezzo
Descrizione
- The economical solution for moving large volume containers around.
- Stainless steel wheels and fastenings.
- Stainless steel recommended in humid environments (no damage to bearings even in the presence of water).
- Load capacity for information purposes only (depending on how the load is spread and the operating temperature).
- Bottom with gaskets on the axle for perfect sealing.
- Volume in litres is equivalent to the container filled to the brim.
Specifica
Specifica
| Tipo | Storage Tote |
| Materiale | HDPE |
| Colore | White |
| Forma | Rectangular |
| Capacità (sistema imperiale) | 81.9 gal. |
| Capacità (sistema metrico) | 310 L |
| Include | 4 Pivoting Wheels, 2 Brakes |