Learn More
Lovibond™ LC 100 Hand-Held Imaging Spectrophotometer
Creates conformance reports in either PDF or Excel formats that can easily be shared with PC or compatible printer
Marca:  Lovibond™ 401120
Dettagli aggiuntivi : Peso : 0.20000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 15360815
  • 4020.00€ / Pezzo
Spedizione stimata: 08-07-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • Each measurement combines 27 images illuminated with different colours and from different directions
  • Enables preview of measurement area and storage and recall of image after measurement
  • Measures and stores up to 20 reference standards with each date and time stamped with option to name
  • Stores up to 350 sample measurements on device and compares to reference standards
  • Samples can be tagged via on-screen keyboard for text or via built in voice recorder
  • For indoor use only
LC 100 and SV 100 Integrated Kit
  • Contents: spectrophotometer, holder, sample cells, USB cord, carrying case
  • Food and Beverages, Textile, Chemicals, Pharmaceutical and Cosmetics, Medical and Clinical
Specifica
Specifica
USB
Food and beverages, coatings, textile, chemicals, pharmaceutical and cosmetics, medical and clinical, household products, Plastics, sugar solutions, syrups and honey
1.8 sec.
TFT
Spectrophotometer, holder, sample cells, USB cord, carrying case
Typical 0.10 ΔE 94 on white (D65/10)