missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LCAT Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA316791
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Suggested positive control: antigen standard for LCAT (transient overexpression lysate), Lipoprotein fraction of whole human plasma.
Lecithin: cholesterol acyltransferase (LCAT) is the plasma enzyme responsible for most HDL cholesteryl esters in human plasma. In species with cholesteryl ester transfer protein (CETP) it is also responsible for a significant proportion of VLDL and LDL cholesteryl esters. LCAT, through the esterification of cholesterol, plays a role in the reverse cholesterol transport pathway by facilitating the net movement of cholesterol from peripheral tissues back to the liver for excretion. LCAT deficiency results in low levels of plasma HDL, whereas a transgenic overexpression of LCAT results in increased plasma HDL concentrations.
Specifica
| LCAT | |
| Polyclonal | |
| Unconjugated | |
| LCAT | |
| AI046659; D8Wsu61e; Lcat; lecithin cholesterol acyltransferase; lecithin-cholesterol acyltransferase; lecithin-cholesterol acyltransferase Lcat; phosphatidylcholine-sterol acyltransferase; phosphatidylcholine--sterol O-acyltransferase; Phospholipid-cholesterol acyltransferase; testicular secretory protein Li 24 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| Conc. Not Determined | |
| Whole serum with 0.05% sodium azide | |
| P04180, P16301 | |
| LCAT | |
| Carboxyl-terminal histidine-tagged human recombinant LCAT purified from the medium of CHO cells overexpressing human LCAT cDNA. | |
| RUO | |
| 16816, 3931 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto