Learn More
DiscoverX LCK Inhibitor
Codice prodotto. 17392629
Encompass
Click to view available options
:
1 mg
Codice prodotto. 17392629
Marca: DiscoverX 921322
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
LCK Inhibitor is an inhibitor of LCK kinases. Provided as 1mg of dry powder.
LCK (lymphocyte-specific protein tyrosine kinase) inhibitor is a pyrrolopyrimidine that blocks the activity of two forms of Lck kinase, Lck (64-509) and Lckcd, and has been shown to inhibit T cell receptor-stimulated IL-2 production. The LCK protein is a member of the Src-family of non-receptor protein tyrosine kinases and plays a critical role in the initial steps of T cell receptor signaling that trigger the production of cytokines. Provided as 1mg of dry powder.
Specifica
Quantità 1 mg