missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LCOR Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA584354
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: NSEEGNTCII PQRNLFKALS EEAWNSGFMG NSSRTADKEN TLQCPKTPLR QDLEANEQDA RPKQENHLHS LGRNKVGYHL HPSDKGQFD Highest antigen sequence indentity to the following orthologs: Rat - 70%, Mouse - 72%.
LCOR is a transcriptional corepressor widely expressed in fetal and adult tissues that is recruited to agonist-bound nuclear receptors through a single LxxLL motif, also referred to as a nuclear receptor (NR) box.
Specifica
| LCOR | |
| Polyclonal | |
| Unconjugated | |
| LCOR | |
| 3110023F06Rik; A630025C20Rik; C10orf12; KIAA1795; Lcor; ligand dependent nuclear receptor corepressor; ligand-dependent corepressor; Mblk1-related protein 2; Mblk1-related protein-2; mKIAA1795; MLR2; RGD1311704; transcription factor MLR2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84458 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96JN0 | |
| LCOR | |
| Recombinant protein corresponding to Human LCOR. Recombinant protein control fragment (Product #RP-108426). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto