Learn More
Bohlender™ Lemo Compact PTFE Double Temperature Probes
Codice prodotto. p-7158921
Click to view available options
Lunghezza (sistema metrico):
400 mm (Total), 300 mm (Usable)
600 mm (Total), 500 mm (Usable)
Dimensione della confezione:
Pezzo
Codice prodotto. 10394174
Marca: Bohlender™ P174020
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
For parallel temperature measurement in aggressive liquids, these probes provide double safety due to redundant systems.
  • Providing temperature resistance from −200° to +250°C
  • Universal chemical resistance
  • Measuring range from −50° to +250°C
  • Two independent measuring sensors in one PTFE-encapsulated stainless steel tube
  • Collar ring diameter of 12 mm
  • Connection by two couplings fixed directly at the end of the probe
  • Typical response times of T 50: 20 to 24 seconds and T 90: 30 seconds
  • Control function due to two independent measuring sensors
  • Suitable for simultaneous temperature measurement and safety circuit as per the specifications of the standard DIN EN 61010-2-010 but only NS/GL socket will be occupied, separately switched sensors
  • Ideal for built-in measurement cables
Specifica
Tipo Probe, Double Temperature
Certificazione/Conformità CE marked
Materiale PTFE
Da utilizzare con (applicazione) Parallel temperature measurement in aggressive liquids
Tempo di risposta T 50: 20 to 24 s; T 90: 30 s
Temperature Range (Metric) -50°C to +250°C
Lunghezza (sistema metrico) 400 mm (Total), 300 mm (Usable)