Learn More
Emma Safety Footwear Leo All-Round Safety D-Fit Shoes
Click to view available options
Size:
40
41
42
43
44
45
46
47
Dimensione della confezione:
1 paio
Descrizione
- Full grain leather uppers
- Hydro-Tec™ sanitized silver lining
- Non-woven antistatic insole
- GripForce™ Easy Twist outsole
- Strobel-stitched upper construction
Specifica
Specifica
| Colore | Black |
| Materiale rivestimento interno | Hydro-Tec Sanitized Silver lining |
| Materiale | Leather |
| Size | 47 |
| Caratteristiche suola | Non-woven antistatic insole |
| Materiale suola | GripForce™ Easy Twist outsole |
| Materiale dello zoccolo | Steel |
| Product Type | All-Round Safety Shoes |