Learn More
Greiner Bio-One Leucosep Polypropylene Centrifugation Tubes
Click to view available options
Capacità (sistema metrico):
12 mL
50 mL
Sterilità:
Aseptically Manufactured
Sterile
Forma del fondo:
Conical
Round
Dimensione della confezione:
pezzi 250
pezzi 500
Descrizione
- Enrichment directly from whole blood
- Simplified filling through porous barrier
- Rapid separation in 15 minutes at room temperature
- Removal of erythrocytes and granulocytes
- No recontamination with erythrocytes
-
Service Plan
No
Specifica
Specifica
| Capacità (sistema metrico) | 12 mL |
| Materiale | Polypropylene |
| Da utilizzare con | Separation of lymphocytes and mononuclear cells from peripheral blood and bone marrow |
| Include | Porous barrier |
| Sterilità | Aseptically Manufactured |
| Forma del fondo | Round |
| Product Type | Centrifuge Tube (Pre-filled) |