Learn More
LIFR Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 22779-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Signal-transducing molecule. May have a common pathway with IL6ST. The soluble form inhibits the biological activity of LIF by blocking its binding to receptors on target cells.Specifica
| LIFR | |
| Polyclonal | |
| Unconjugated | |
| LIFR | |
| CD118, LIF R, LIF receptor, LIFR, SJS2, STWS, SWS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16880, 3977 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P42702, P42703 | |
| LIFR | |
| LIFR Fusion Protein Ag18742 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |