Learn More
Cole-Parmer™ Lightnin Heavy-Duty Gear Drive Mixer
Codice prodotto. 15467453 Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Speed:
20 to 280 rpm
20 to 350 rpm
Modello:
EV5P25A1D36
EV5P25A1F48
EV5P25A1J60
EV5P25ET1F48
EV5P25M1E36
EV5P25M1F48
EV5P25M1J60
EV5P25T1F48
EV5P25T1J60
EV5P50A2L36
EV5P50M2L36
EV5P50T2L36
EV5P5OAIP60
EV6P75M2X60
EV6P75T2T48
EV6P75T2X60
Tipo di motore:
Air-Driven
TEFC
TENV
XPRF
Coppia:
1440 in. oz.
2701 in. oz.
720 in. oz.
Dimensione della confezione:
Pezzo
Codice prodotto. 15467453
Marca: Cole-Parmer™ 5031046
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Lightnin Heavy-Duty Gear Drive Mixers are ideal for mixing your high-viscosity materials or large batches of water-like fluids. All models maintain speeds from 20 to 350 rpm (20:1 ratio) and generate enough torque to mix highly viscous fluids.
  • Includes:
    • 316 SS shaft, 316 SS A-310 impeller, and drive quill with set screw.
    • Models 50310-68, -70 and -72 also include bung adapter and a folding impeller.
  • More About This Item:
    • These variable-speed, gear-drive mixers are ideal for mixing your high-viscosity materials or large batches of water-like fluids. All models maintain speeds from 20 to 350 rpm (20:1 ratio) and generate enough torque to mix highly viscous fluids.
    • Gear-drive mixers mount with 20°> offset tank clamp (included). Available as either an air-powered or electric mixer.
    • Select from three speed options; 20 to 350 rpm (20:1 ratio), 20 to 280 rpm (14:1 ratio), or fixed speed of 350 or 280 rpm. The 3⁄4-hp mixers generate plenty of torque for your high-viscosity applications.
Specifica
Speed 20 to 350 rpm
Diametro (sistema imperiale) albero 0.75 in.
Diametro (sistema metrico) albero 19.05 mm
Include 316 SS Shaft, 316 SS A-310 Impeller, and Drive Quill with Set Screw
Materiale 316 Stainless Steel
Modello EV5P25ET1F48
Tipo di motore XPRF
Coppia 720 in. oz.