Learn More
Cole-Parmer™ Lightnin Heavy-Duty Gear Drive Mixer
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Speed:
20 to 280 rpm
20 to 350 rpm
Modello:
EV5P25A1D36
EV5P25A1F48
EV5P25A1J60
EV5P25ET1F48
EV5P25M1E36
EV5P25M1F48
EV5P25M1J60
EV5P25T1F48
EV5P25T1J60
EV5P50A2L36
EV5P50M2L36
EV5P50T2L36
EV5P5OAIP60
EV6P75M2X60
EV6P75T2T48
EV6P75T2X60
Tipo di motore:
Air-Driven
TEFC
TENV
XPRF
Coppia:
1440 in. oz.
2701 in. oz.
720 in. oz.
missing translation for 'unitSize'
Pezzo
Description
- Includes:
- 316 SS shaft, 316 SS A-310 impeller, and drive quill with set screw.
- Models 50310-68, -70 and -72 also include bung adapter and a folding impeller.
- More About This Item:
- These variable-speed, gear-drive mixers are ideal for mixing your high-viscosity materials or large batches of water-like fluids. All models maintain speeds from 20 to 350 rpm (20:1 ratio) and generate enough torque to mix highly viscous fluids.
- Gear-drive mixers mount with 20°> offset tank clamp (included). Available as either an air-powered or electric mixer.
- Select from three speed options; 20 to 350 rpm (20:1 ratio), 20 to 280 rpm (14:1 ratio), or fixed speed of 350 or 280 rpm. The 3⁄4-hp mixers generate plenty of torque for your high-viscosity applications.
Specifications
Specifications
| Speed | 20 to 350 rpm |
| Diametro (sistema imperiale) albero | 0.75 in. |
| Diametro (sistema metrico) albero | 19.05 mm |
| Cavalli motore | 1/2 hp |
| Include | 316 SS Shaft, 316 SS A-310 Impeller, and Drive Quill with Set Screw |
| Materiale | 316 Stainless Steel |
| Modello | EV5P50A2L36 |
| Tipo di motore | Air-Driven |
| Coppia | 1440 in. oz. |