Learn More
Novus Biologicals™ Lightning-Link (R) AMCA Antibody Labeling Kit
Codice prodotto. 18045416 Sfoglia Tutto Bio Techne Prodotti
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µg
Codice prodotto. 18045416
Marca: Novus Biologicals™ 3130005
Antibody labeling kit
Lightning-Link™ Rapid is an innovative technology that enables direct labeling of proteins, peptides or other biomolecules for use in R&D applications, drug discovery and the development of diagnostic kits
Kit Contents:
  • 1 vial of LL Rapid Modifier reagent
  • 1 vial of LL Rapid Quencher reagent
  • 1 glass vial of Lightning-Link™ Rapid mix
TRUSTED_SUSTAINABILITY
Specifica
Quantità 100 μg
Product Type Antibody Labeling Kit
Content And Storage -20°C