Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Atto565 Antibody Labeling Kit
Sfoglia Tutto Bio Techne Prodotti
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µg
Descrizione
Lightning-Link™ Rapid Antibody Labeling kits enable direct labeling of antibodies, proteins, peptides or other biomolecules and allow conjugations to be set up in seconds, and used within minutes (less than 20 minutes).
Kit Contents:
- Lightning-Link™ Rapid mix
- 1 vial of LL Rapid Quencher reagent
- 1 vial of LL Rapid Modifier reagent
Specifica
Specifica
| Quantità | 100 μg |
| Product Type | Antibody Labeling Kit |
| Content And Storage | -20°C |