Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Atto633 Antibody Labeling Kit
Codice prodotto. 18030514 Sfoglia Tutto Bio Techne Prodotti
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µg
Codice prodotto. 18030514
Marca: Novus Biologicals™ 3530005
Antibody labeling kit
Lightning-Link™ Rapid Antibody Labeling kits enable direct labeling of antibodies, proteins, peptides or other biomolecules and allow conjugations to be set up in seconds, and used within minutes (less than 20 minutes).
Kit Contents:
  • Lightning-Link™ Rapid mix
  • 1 vial of LL Rapid Quencher reagent
  • 1 vial of LL Rapid Modifier reagent
TRUSTED_SUSTAINABILITY
Specifica
Quantità 100 μg
Product Type Antibody Labeling Kit
Content And Storage -20°C