Learn More
LIMK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12350-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Proteins containing LIM motifs are typically involved in cell fate determination and growth control. A family of proteins designated LIM kinases, including LIMK-1 and LIMK-2, has been identified. LIMK-1 has been shown to regulate the stabilization of F-Actin structures and cofilin activity, indicating that LIMK-1 plays a role in a signaling pathway involved in the regulation of cell motility and morphogenesis. LIMK-1 inhibits neuronal differentiation of PC12 cells, and is thought to act by interfering with events downstream of MAPK activation. Expression patterns of LIMK-1 and LIMK-2 suggest that these proteins may have different functions during development. A truncated form of LIMK-2 has been identified in adult testis that is thought to arise from an alternative initiation exon.Specifica
| LIMK2 | |
| Polyclonal | |
| Unconjugated | |
| LIMK2 | |
| LIM domain kinase 2, LIMK 2, LIMK2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16886, 29524, 3985 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O54785, P53670, P53671 | |
| LIMK2 | |
| LIMK2 Fusion Protein Ag3011 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |