Learn More
Agilent BioTek Liquid Handling Control Software
Sfoglia Tutto Agilent BioTek Prodotti
Click to view available options
Include:
E-delivery, 5 Installation Licenses
E-delivery, 5 Installation Licenses, 21CFR Part 11 Compliance
Dimensione della confezione:
Pezzo
Descrizione
- LHC software makes it easy to program, even the most complex protocols with a choice of deployment
- Protocols can be run directly from the computer or downloaded to the instrument for convenient onboard recall and operation
- Full instrument control
- For use with 405 TS Washer, 405 LS Washer, ELx405 Select Deep Well Washer, 50 TS Washer, MultiFlo Dispenser, MultiFlo FX Dispenser, EL406 Washer Dispenser, 406 FX Washer Dispenser
- Batch processing with connected BioStack Microplate Stacker or BioSpa 8 Automated Incubator
- Seamless robotic integration to commercially available robotic systems
Specifica
Specifica
| Include | E-delivery, 5 Installation Licenses |
| Da utilizzare con (apparecchiatura) | Agilent BioTek Microplate Washers, Dispensers, and Liquid Handling Systems |
| Tipo | Liquid Handling Control Software |