Learn More
Lithium Dodecylsulfate, ≥98%
Product Code. 15865208 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15865208 25 g Plastic bottle 25g
15875208 100 g Plastic bottle 100g
15865358 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15865208 Supplier Thermo Scientific Supplier No. J32816.14
 Restricted Item
In stock
Add to basket
CAS: 2044-56-6 | C12H25LiO4S | 272.33 g/mol
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2044-56-6
Molecular Formula C12H25LiO4S
Molecular Weight (g/mol) 272.33
MDL Number MFCD00007467
InChI Key YFVGRULMIQXYNE-UHFFFAOYSA-M
Synonym lithium dodecyl sulfate, lithium dodecylsulfate, sulfuric acid, monododecyl ester, lithium salt, lauryl sulfate lithium salt, lithium lauryl sulfate, dodecyl lithium sulfate, lds, lithium 1+ ion dodecyl sulfate, sulfuric acid, monododecyl ester, lithium salt 1:1, lithium dodecyl sulfate, ultra pure
PubChem CID 2735071
SMILES [Li+].CCCCCCCCCCCCOS([O-])(=O)=O
Specifications
Physical Form Powder
Quantity 25 g
Packaging Plastic bottle
Linear Formula CH3(CH2)11OSO3Li
UN Number UN1325
IUPAC Name lithium;dodecyl sulfate
Formula Weight 272.33
Percent Purity ≥98%
Chemical Name or Material Lithium Dodecylsulfate