Learn More
Lithium octanoate, 99%
Click to view available options
Quantità:
25 g
100 g
500 g
Dimensione della confezione:
100g
25g
500g
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Materiale o nome chimico | Lithium octanoate |
| Quantità | 500 g |
| Peso formulazione | 150.14 |
| Percent Purity | 99% |
| Forma fisica | Powder |