Learn More
Brannan™ LO-tox™ Lab Thermometer with 3-points Traceable Calibration
Codice prodotto. 17853018
Click to view available options
Temperature Range (Metric):
-1°C to 101°C
-1°C to 51°C
-10°C to +110°C
-10°C to +210°C
-10°C to +260°C
-10°C to +300°C
-10°C to +330°C
-10°C to +360°C
-10°C to +50°C
-20°C to +110°C
-20°C to +150°C
-35°C to +50°C
Suddivisione:
0.1°C
0.2°C
0.5°C
1°C
2°C
Tipo di immersione:
Partial Immersion
Total Immersion
Lunghezza (sistema metrico):
15.5 cm
30.5 cm
40.5 cm
Dimensione della confezione:
Pezzo
Codice prodotto. 17853018
Marca: Brannan™ 44/801/8 + 49/101/0
LO-tox™ lab thermometer complete with calibration certificate.
  • LO-tox™ is an organic filling specially chosen to achieve a wide temperature span and provide the lowest practical levels of hazard
  • Safe non-mercury filling
  • Supplied with calibration certificate
  • Non-roll end cap
  • Supplied as standard in protective individual, resealable telescopic tube
  • Made in UK
Specifica
Punti di calibrazione 0°C, 20°C, 35°C
Certificazione/Conformità UK Traceable to National Standards ISO 9001
Tipo di immersione Partial Immersion
Lunghezza (sistema metrico) 30.5 cm
Materiale Glass
Riempimento termometro Blue Spirit
Max. Temperature (English) 122°F
Temperatura max (sistema metrico) +50°C
Min. Temperature (English) -31°F
Min. Temperature (Metric) -35°C
Intervallo di temperatura (sistema imperiale) -31°F to +122°F
Temperature Range (Metric) -35°C to +50°C
Accuracy ± 1.0°C at 35°C
Suddivisione 1°C
Immersion Length 76 mm
Larghezza (sistema metrico) 1.2 cm