Learn More
Fisherbrand™ Logger-Trac™ Mini-Logger-Trac Datalogging Thermometer
Sfoglia Tutto Fisherbrand ProdottiDescrizione
Requires
Requires MaxiThermal software (15-081-150) and cradle with USB cable (14-650-123) to download data and restart mini datalogger.
Know the temperature excursion duration immediately, no data download required with Logger-Trac™ Mini-Logger-Trac Datalogging Thermometer.
- Duration of Temperature excursion
- Data-logger with user-defined recording intervals
- Active LCD Display shows the following important data: Current temperature, Minimum/Maximum temperatures, Recording status, Run Time, Alarm (Excursion) Cumulative Time, Alarm Tally
- Requires the Fisherbrand Traceable Cradle and USB Cable (6549) to upload data to PC
USER SELECTABLE RECORDING INTERVALS For complete flexibility, the user-defined recording interval may be set based on the number of desired days to log (from 4 hours to 365 days) or the desired time interval (from every 2 seconds to every 24 hours).Internal flash memory stores 7680 readings.
To ensure that no data is lost, once the memory is full the thermometer will stop logging data (data will not be overwritten).While recording, the following information is displayed: Current, Minimum and Maximum temperature achieved and cumulative running time. Once an alarm condition has occurred, the following additional information is displayed: Alarm Tally, ALARM notification, cumulative alarm time.
Individual Logger-Trac Thermometers are pre-programmed with alarm values of 2 °C (low) and 8 °C (high) and a recording interval to allow for 30 days of recording (readings are logged every 5 minutes, 33 seconds).
Specifica
Specifica
| Tipo | Datalogger |
| Tipo di display | LCD |
| Memoria dati | 7680 Readings |
| Calibrazione | Not Calibrated |
| Tipi di allarme | High/Low |
| Software Type | TraceableLIVE |
| Descrizione | With temperature MIN/MAX memory. data logging and exportable data |
| Interfaccia | USB |
| Protezione IP | IP66 |
| Tipi di sensore | Internal Sensor |
| Vedi altri risultati |