Learn More
Lovibond™ Reactive Phosphorus Orthophosphate Reagent Set
Click to view available options
N. di test:
50
Dimensione della confezione:
pezzi 50
Descrizione
Includes
Phosphate 3 Reagent (10mL sample, 50 pack), Dilution Tube (50 ct.), Deionized water (100mL)
- Offers same high quality standards as tablet reagents
- Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
- Simply tear open Powder Pack and pour reagent into sample
- Large design makes them easier to handle
- “Easy Open” notch makes it simple to tear open package
- Easily create “V”, to allow user to pour reagent into sample
- Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifica
Specifica
| Da utilizzare con (applicazione) | Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing |
| Da utilizzare con (apparecchiatura) | Hach™ and other manufacturers test kits, colorimeters and spectrophotometers |
| Include | 10mL phosphate 3 reagent (50 pack), 50 ct. dilution tube, 100mL deionized water |
| N. di test | 50 |
| Range | 0.06 to 5mg/L |
| Metodo test | Ascorbic Acid |
| Tipo | Reactive Phosphorous Reagent Set |