Learn More
Lovibond™ Replacement Reagent for CHECKIT™ Comparator
Codice prodotto. 15368970
Click to view available options
Quantità:
100 Pack
100 mL
15 mL
200 Pack
250 Pack
Tipo:
DEHA Test Solution
DPD Glycine Test Tablets
DPD Glycine Test Tablets (for Cl separation)
EDTA Test Tablets
pH-Universal
Formato:
Liquid
Tablet
Dimensione della confezione:
100 pastiglie
100mL
15mL
Pezzo
pezzi 100
pezzi 250
Codice prodotto. 15368970
Marca: Lovibond™ 512171BT
Lovibond™ Replacement Reagent for CHECKIT™ Comparator is manufactured under tightly controlled conditions to maintain the highest quality standards.
  • Highest quality production standards
  • Consistent and reliable reagents are provided in liquid and tablet formats
  • Maximum accuracy
Specifica
Formato Tablet
Da utilizzare con (applicazione) Water analysis
Da utilizzare con (apparecchiatura) Colorimeter and spectrophotometer
Quantità 200 Pack
Tipo DPD Glycine Test Tablets