missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRP4 Monoclonal Antibody (741704)
Mouse Monoclonal Antibody
Marca: Invitrogen™ MA524353
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
This gene encodes a member of the low-density lipoprotein receptor-related protein family. The encoded protein may be a regulator of Wnt signaling. Mutations in this gene are associated with Cenani-Lenz syndrome.
Specifica
| LRP4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O75096 | |
| LRP4 | |
| E. coli-derived recombinant human LRP-4 Gly18-Ser344. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 741704 | |
| Unconjugated | |
| LRP4 | |
| 6430526J12Rik; CLSS; CMS17; Corin; D230026E03; Kiaa0816; LDL receptor related protein 4; LDLR; LDLR dan; low density lipoprotein receptor-related protein 4; low-density lipoprotein receptor-related protein 4; LRP10; Lrp4; LRP-4; mdig; Megf7; multiple epidermal growth factor-like domains 7; SOST2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 4038 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Spot an opportunity for improvement?Share a Content Correction