Learn More
LRR 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11628-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
May negatively regulate the 4-1BB-mediated signaling cascades which result in the activation of NK-kappaB and JNK1. Probable substrate recognition subunit of an ECS (Elongin BC- CUL2/5-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.Specifica
| LRR 1 | |
| Polyclonal | |
| Unconjugated | |
| LRR1 | |
| 4 1BBLRR, LRR 1, LRR repeat protein 1, LRR1, PPIL5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 122769, 214345, 685860 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q80VQ1, Q96L50 | |
| Lrr1, Lrrc1 | |
| LRR 1 Fusion Protein Ag2159 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |