Learn More
Merck Substrato Western HRP Chemiluminescence Immobilon
Codice prodotto. p-3981884 Sfoglia Tutto Merck Prodotti
Click to view available options
Quantità:
500 ml
Product Type:
Substrato Luminata Classico Western HRP
Substrato Luminata Crescendo Western HRP
Substrato Luminata Forte Western HRP
Sensibilità:
∽400 fg
∽6 pg
Da ∽1 a 3 pg
Dimensione della confezione:
500mL
Pezzo
Codice prodotto. 12634645
Marca: Merck WBLUF0500
Premixed, ready-to-use chemiluminescent reagents for detection of HRP-based Westerns
Horseradish peroxidase (HRP) catalyzes the oxidation of luminol in the presence of peroxide, releasing photons (chemiluminescence) that are captured on x-ray films or by CCD cameras. Most HRP substrates are plagued by quickly decaying signal and short shelf life, but Luminata substrates are formulated differently to include stabilizing reagents.
  • Consistent results with less pipetting error
The stabilizers added to Luminata substrates:
  • Prevent spontaneous oxidation of luminol by peroxide in the absence of HRP
  • Enhance the stability of both the luminol and peroxide, ensuring that Luminata substrates are stable for one year in their premixed format
  • Stabilize, strengthen, and prolong the chemiluminescence signal once HRP starts to catalyze oxidation
Three different sensitivities for a range of detection needs:
  • Immobilon Classico premixed reagent for everyday detection needs
  • Immobilon Crescendo premixed reagent for detection in the picogram range
  • Immobilon Forte premixed reagent for detection in the femtogram range
TRUSTED_SUSTAINABILITY
Specifica
Product Type Substrato Luminata Forte Western HRP
Da utilizzare con (applicazione) Dot/Slot Blot
Quantità 500 ml
Sensibilità ∽400 fg
Content And Storage da 18° a 24 °C