Learn More
DiscoverX Luteinizing Hormone Releasing Hormone (LH-RH), Human
Click to view available options
Quantità:
5 mg
Descrizione
Luteinizing Hormone Releasing Hormone (LH-RH, GnRH) is a hypothalamic peptide hormone that stimulates the production and release of follicle-stimulationg hormone (FSH) and luteinizing hormone (LH) from the anterior pituitary, subsequently initiating and maintainining reproductive organ functions. Agonist of the gonadotropin-releasing hormone receptor (GnRHR). Provided as 5mg of dry powder alongside reconstitution buffer.
Specifica
Specifica
| Product Type | Control Ligands |
| Quantità | 5 mg |