missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LXR beta/NR1H2 Antibody (LXRB/2731), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-08914F
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
LXR beta/NR1H2 Monoclonal antibody specifically detects LXR beta/NR1H2 in Human samples. It is validated for ELISA, Protein Array
Specifica
| LXR beta/NR1H2 | |
| Monoclonal | |
| FITC | |
| Liver X receptor beta, LXR-b, NER-I, NERliver X receptor-beta, nuclear orphan receptor LXR-beta, Nuclear receptor NER, Nuclear receptor subfamily 1 group H member 2, nuclear receptor subfamily 1, group H, member 2, oxysterols receptor LXR-beta, RIP15, steroid hormone-nuclear receptor NER, Ubiquitously-expressed nuclear receptorLXRB, UNRLX receptor beta | |
| Recombinant full-length human LXR beta/NR1H2 protein (Uniprot: P55055) | |
| 100 μg | |
| Diabetes Research, Lipid and Metabolism | |
| 7376 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
| ELISA, Protein Array | |
| LXRB/2731 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto