Learn More
Labconco™ Lyph-Lock™ Freeze-Dry Flask Assembly
Sfoglia Tutto Labconco Corporation Prodotti
Click to view available options
Capacità (sistema metrico):
50 mL
100 mL
250 mL
500 mL
750 mL
1000 mL
Dimensione della confezione:
Pezzo
Descrizione
Complete three-piece flask consists of high-strength borosilicate glass molded top and bottom with silicone rubber seal
- With 24/40 joints
- Seal grasps top and bottom securely—never comes in contact with sample
- Flat bottomed
- Wide mouth lets you load samples easily and lyophilize quickly
- Compatible with Labconco, Virtis™ and other major brands of freeze dryers
- Flasks should be filled to no more than one-third the volume so that maximum surface area is achieved for efficient lyophilization
Required Accessories
- Adapters are required to attach flasks to freeze dry valve ports
Specifica
Specifica
| Diametro (sistema imperiale) esterno | 1.33 in. |
| Capacità (sistema metrico) | 50 mL |
| Materiale | Borosilicate Glass |
| Altezza (sistema imperiale) | 2.63 in. |
| Altezza (sistema metrico) | 6.7 cm |
| Include | Complete three-piece flask consists of high-strength Borosilicate Glass molded top and bottom with Silicone Rubber seal |
| Product Type | Flask Kit |
| Giunto rastremato standard | 24/40 |
| Peso (sistema imperiale) | 3 lb. |
| Monouso | Reusable |
| Vedi altri risultati |