Learn More
MP Biomedicals™ TeenPrep™ Lysing Matrix C, 15 mL Tubes
Sfoglia Tutto MP Biomedicals Prodotti
Click to view available options
Quantità:
25 x 15 mL Tubes
5 x 15 mL Tubes
50 x 15 mL Tubes
Dimensione della confezione:
750mL
pezzi 25
pezzi 5
Descrizione
Lysing Matrix C contains 1 mm silica spheres and can be used with FastPrep instruments or similar bead beater.
- Tube Size: 15 mL
- Sample Type: Yeast, fungi, mold, spores and filamentous algae
- Bead Type: 1 mm silica spheres
Specifica
Specifica
| Tipo di profilo | Silica Spheres |
| Content And Storage | Store at Room Temperature. |
| Tipo di campione | Yeast, Fungi, Algae |
| DNasi | Dnase-free |
| Da utilizzare con (apparecchiatura) | FastPrep instruments or other bead beaters |
| Da utilizzare con (applicazione) | Nucleic Acid Isolation and Purification, Homogenization |
| Matrix | Lysing Matrix C |
| Quantità | 25 x 15 mL Tubes |
| RNasi | RNase-free |
| Volume del campione | Up to 2 g per prep |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.