Learn More
Thermo Scientific™ M110 Muffle Furnaces
Heat and dry metals at high temperatures with Thermo Scientific™ M110 Muffle Furnaces, that evenly distribute heat and operate economically in a small footprint.
dietro preventivo
Specifica
Certificazione/Conformità | CE mark |
---|---|
Capacità (sistema metrico) | 9 L |
Temperatura (sistema metrico) | 1100°C |
Consumo energetico | 2.9 kw |
Uniformità termica | ±7°K |
Descrizione
Heat and dry metals at high temperatures with Thermo Scientific™ M110 Muffle Furnaces, that evenly distribute heat and operate economically in a small footprint.
Key Features:
- 9L working volume
- Two-shell design with air pocket between annealing box and outside walls
- Multiple layers of high-quality, asbestos-free ceramic fiber insulation
- Heating elements safely positioned in grooved blocks on side walls of the work space, covered with ceramic plates
- Hinged door designed with parallel forced guidance system—hot surface always faces away from user when door is open
- Adjustable upper limit cut-out protects samples and equipment
Ordering Information:
Models available with optional 24 hr. timer, Thermo Scientific™ Thermicon™ P digital program controller with microprocessor, exhaust fan and/or exhaust flue. Additional exhaust flue (50006394) and tray (50006408) available separately. Not available in North America.
Compatible with:
- Exhaust flue (50006394)
- Tray (50006408)
Specifica
CE mark | |
1100°C | |
±7°K | |
28.4 x 22.7 x 29.6 in. (72 x 57.6 x 75.2 cm) | |
72 cm | |
57.6 cm | |
29.6 in. | |
75.2 cm | |
11.8 in. | |
172 lb. | |
230 V 50/60 Hz | |
50/60 Hz |
9 L | |
2.9 kw | |
11.8 x 7.9 x 5.9 in. (30 x 20 x 15 cm) | |
28.4 in. | |
7.9 in. | |
20 cm | |
5.9 in. | |
15 cm | |
30 cm | |
78 kg | |
230 V |