Learn More
uvex MACSOLE™ Boots S7
Sfoglia Tutto Uvex Prodotti
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Dimensione della confezione:
1 paio
Descrizione
The uvex 3, but even more robust.
- The MACSOLE™ S7 68633 boots with rubber outsole provides even stronger support for heavy-duty applications, making it particularly resistant to heat, chemicals and cuts.
- The MACSOLE™ rubber sole technology also impresses with its unbeatable slip resistance (SR), even in the cold.
- EN ISO 20345: S7
Specifica
Specifica
| Colore | Black, Blue |
| Materiale | Rubber, Leather |
| Size | 43 |
| Materiale dello zoccolo | Polymer |
| Monouso | Reusable |
| Sesso | Men's |
| Fit | Wide Width |
| Senza lattice | No |
| Waterproof | Yes |
| Product Type | Boots |