Learn More
MAGEA1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12233-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Human maligt neoplasms carry rejection antigens that are recognized by the patients′ autologous, tumor directed and specific, cytolytic, CD8+ T lymphocyte clones (CTL). An important group of antigens is coded by the MAGE family of genes. It was identified that melanoma associated with antigens (MAAs). Because MAGE-1 is expressed on a significant proportion of human neoplasms of various hisological tyoes (melanoma, brain tumors of glial origin, neuroblastoma, non-small cell lung cancer, breast, gastric, colorectal, ovarian, renal cell carcinomas) and not on normal tissues, the encoded antigen may serve as a marker of early detection and target for cancer immunotherapy.Specifica
| MAGEA1 | |
| Polyclonal | |
| Unconjugated | |
| MAGEA1 | |
| Antigen MZ2 E, Cancer/testis antigen 1.1, CT1.1, MAGE 1 antigen, MAGE1, MAGE1A, MAGEA1, Melanoma associated antigen 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4100 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P43355 | |
| MAGEA1 | |
| MAGEA1 Fusion Protein Ag2871 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |