Learn More
Hanna Instruments™ Ammonia Reagent Sets, For Colorimetric Water Testing
Click to view available options
Da utilizzare con (applicazione):
Ammonia High Range
Magnesium and Total Hardness
N. di test:
25
100
Dimensione della confezione:
100 tests
pezzi 100
pezzi 25
Descrizione
These high quality reagents are designed to follow the ASTM Manual of Water and Environmental Technology, D1426, Nessler Method for colorimetric determination of Ammonia. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
- Pre-made, pre-measured testing reagents for ease of use
- Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
- Clearly marked with expiration date and lot number on each component for traceability
- Tests available for low-range, medium range and high-range determinations
- Kits with 25 ready-to-use single vials of reagent available
Specifica
Specifica
| Da utilizzare con (applicazione) | Ammonia High Range |
| Da utilizzare con (apparecchiatura) | Compatible Hanna benchtop and portable photometers |
| Include | Dropper bottles |
| N. di test | 100 |
| Quantità | 1 Set |
| Range | 0.0 to 50 ppm Ammonium |
| Metodo test | Cholorimetric determination |
| Tipo | Water Testing Kit |