Learn More
Value Plastics Male Luer with Integral Lock Ring to 400 Series Barb
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Diametro (sistema imperiale) interno:
1/16 in.
3/32 in.
Materiale:
Nylon
Polycarbonate
Polypropylene
Dimensione della confezione:
pezzi 25
Descrizione
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifica
Specifica
| Certificazione/Conformità | Non-DEHP;Non-Phthalate;RoHS;FDA |
| Diametro (sistema imperiale) interno | 1/16 in. |
| Materiale | Polycarbonate |
| Ingresso | Male Luer x Hose Barb |
Always test the application to verify the performance of the interconnection.