Learn More
Value Plastics Male Luer Integral Lock Ring to Plug Adapters
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Materiale (guarnizione):
Nylon
Polycarbonate
polypropylene
Dimensione della confezione:
pezzi 25
Descrizione
Luer fittings allow for a simple yet secure twist-lock connection. Ideal for capping off fluid transfer system. Prevent leaks with integral lock ring mechanism
Specifica
Specifica
| Certificazione/Conformità | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Ingresso | Male Luer Integral Lock Ring, Plug, Closed at Luer Tip |
| Materiale (guarnizione) | polypropylene |