Learn More
Hamilton™ Male Luer Lock / Threaded Connector (1/8 - NPT)
Codice prodotto. 11577634
Codice prodotto. 11577634
Marca: Hamilton™ 86536
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Inert fittings and connectors made from CTFE and designed to adapt to and from a variety of male and female luer and threaded connections.
Specification Sheet
  • Connection 1 – Male Luer Lock
  • Connection 2 – Male 1/8 NPT
  • I.D. – 1.5 mm
Specifica
Tipo Male Luer Lock