Learn More
Value Plastics Male Luer Slip to 200 Series Barb
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Diametro (sistema imperiale) interno:
1/16 in.
1/8 in.
3/32 in.
Materiale (guarnizione):
Nylon
Polypropylene
Dimensione della confezione:
pezzi 25
Descrizione
Quick and easy assembly and disassembly without the need for tools. Barbed ridges provide secure grip on the inside of flexible tubing. Slip luer fittings allow for a simple yet secure connection
Specifica
Specifica
| Certificazione/Conformità | Non-DEHP;Non-Phthalate;RoHS |
| Diametro (sistema imperiale) interno | 1/16 in. |
| Ingresso | Male Luer Slip, 200 Series Barb |
| Materiale (guarnizione) | Nylon |