Learn More
Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers
Descrizione
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers, also for use with National or Chromacol vials.
Lightweight, precision crimpers attach seals to chromatography or serum vials. Available for standard aluminum or flip-off seals.
- Factory set to properly crimp standard-thickness seals
- Adjustable handle setting screw allows proper crimps with septa of varying thickness
Manual Decripmers
Forged pliers feature a fitted gripping surface designed to allow removal of the listed sizes of seals.
- Simply grip pliers around top of seal, squeeze and pry upward to remove
Manual Decrimping Pliers
Place decrimping head squarely on seal and squeeze.
- Minimizes damage to vial rim with chromatography or serum vials
- Removes standard aluminum seals and many steel seals
Specifica
Specifica
| Descrizione | Manual Decrimper |
| N. per confezione | 1 Ea. |
| Tipo | Manual Decrimper |
| Da utilizzare con (apparecchiatura) | 11 mm Standard Seal, National Vial, Chromacol Vial |
| Size | 11 mm |
| Da utilizzare con (applicazione) | Industrial Chromatography |
| Linea di prodotti | National |