Learn More
MAP 2a,b,c (Microtubule-Associated Protein) Ab-3 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry experiments with the Epredia™ MAP 2a,b,c (Microtubule-Associated Protein) Ab-3, Mouse Monoclonal Antibody.
190.00€ - 783.00€
Specifica
| Antigene | MAP 2a,b,c (Microtubule-Associated Protein) Ab-3 |
|---|---|
| Clone | AP18 |
| Applicazioni | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classificazione | Monoclonal |
| Coniugato | Unconjugated |
| Codice del prodotto | Marca | Quantità | Forma | Concentrazione | Prezzo | Quantità e disponibilità | |||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Forma | Concentrazione | Prezzo | Quantità e disponibilità | |||
|
12690697
|
Epredia
MS-250-P0 |
100 μL | Concentrated | 200μg/mL | |||||
|
12600707
|
Epredia
MS-250-P1 |
500 μL | Concentrated | 200μg/mL | |||||
|
12680697
|
Epredia
MS-250-P |
1 mL | Concentrated | 200μg/mL | |||||
|
12610707
|
Epredia
MS-250-P1ABX |
100 μL | Concentrated | 1mg/mL | |||||
|
12620707
|
Epredia
MS-250-PABX |
200 μL | Concentrated | 1mg/mL | |||||
|
12650707
|
Epredia
MS-250-R7 |
7 mL | Prediluted | — | |||||
Descrizione
Microtubule Associated Protein 2 exists in two high molecular weight forms (2a & 2b) and a low molecular weight form. The expression of MAP2 is developmentally regulated and its multiple forms arise by alternative splicing of a single gene. Ab-3 is specific to phosphprylated form of MAP2a,b,c and shows no cross-reaction with other MAPs, tau, and tubulin. Ab-3 reacts with dendrites and cell bodies of neurons. It is useful in studies of neuron structure in normal and malignant brain tissue and in Alzheimer's.Host Species: Mouse
Clone: AP18
Isotype: IgG1
Species Reactivity: Human, Cow, Rat, Xenopus, Quail. Others not known.
Epitope: aa 1-151
Immunogen: Bovine brain microtubule associated protein 2
Molecular Weight: ∽280kDa doublet and a 70kDa singlet
Positive Control: IMR-5 or T98G human glioblastoma cells or brain
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Specifica
| MAP 2a,b,c (Microtubule-Associated Protein) Ab-3 | |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| IVD | |
| Bovine, Human, Quail, Rat, Xenopus species | |
| IgG1 |
| AP18 | |
| Monoclonal | |
| Mouse | |
| Neuroscience | |
| Bovine brain microtubule associated protein 2 | |
| Primary |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.