missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2 Antibody (5H11), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP1-92711MFV500
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
MAP2 Monoclonal antibody specifically detects MAP2 in Human, Mouse, Rat, Bovine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry Free-Floating
Specifica
| MAP2 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine, Monkey | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Free Floating) | |
| 5H11 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry - Free Floating | |
| DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2 | |
| MAP2 Antibody (5H11) was developed against full length purified bovine protein, epitope mapped to projection domain of human sequence, amino acids 1057-1588 using MAP2 Projection Domain 3. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers | |
| 4133 | |
| Store at 4C in the dark. | |
| IgG2b |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto