missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPK7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11164-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
ERK5/MAPK7 is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. This kinase is specifically activated by mitogen-activated protein kinase kinase 5. It is involved in the downstream signaling processes of various receptor molecules including receptor type kinases, and G protein-coupled receptors. In response to extracelluar signals, this kinase translocates to cell nucleus, where it regulates gene expression by phosphorylating, and activating different transcription factors. Four alternatively spliced transcript variants of this gene encoding two distinct isoforms have been reported. ERK5 is activated by tyrosine and threonine phosphorylation It is expressed in many adult tissues, abundantly in heart, placenta, lung, kidney and skeletal muscle, but is not detectable in liver.Specifica
| MAPK7 | |
| Polyclonal | |
| Unconjugated | |
| MAPK7 | |
| Big MAP kinase 1, BMK 1, BMK1, ERK 5, ERK4, ERK5, MAP kinase 7, MAPK 7, MAPK7, PRKM7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114509, 23939, 5598 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P0C865, Q13164, Q9WVS8 | |
| MAPK7 | |
| MAPK7 Fusion Protein Ag1648 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto