missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARCKSL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14124-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
MARCKS (Myristoylated alanine-rich protein kinase C substrate), also designated 80K or 80K-L, is a 332 amino acid protein that localizes to the plasma membrane and functions as a major cellular substrate for protein kinase C (PKC). MARCKSL1 (MARCKS-like protein 1), also known as MacMARCKS, MLP, MRP, or F52, is a 195 amino acid protein that, like MARCKS, is a major substrate for PKC. Expressed in a variety of tissues with highest levels found in testis and uterus, MARCKSL1 participates in the coordination of membrane-cytoskeletal signaling events, including secretion, migration, phagocytosis and cell adhesion. Additionally, MARCKSL1 functions as a regulator of Integrin activation and is thought to regulate Integrin-dependent signal transduction pathways, especially those involved in macrophage spreading.Specifica
| MARCKSL1 | |
| Polyclonal | |
| Unconjugated | |
| MARCKSL1 | |
| F52, Mac MARCKS, MACMARCKS, MARCKS like 1, MARCKS like protein 1, MARCKS related protein, MARCKSL1, MLP, MLP1, MRP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 65108 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P49006 | |
| MARCKSL1 | |
| MARCKSL1 Fusion Protein Ag5274 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto