Learn More
Brady™ Marine Labels for BradyJet J4000 and J5000, BS 5609
Click to view available options
Altezza (sistema metrico):
85 mm
110 mm
155 mm
210 mm
279.4 mm
Larghezza (sistema metrico):
55 mm
75 mm
105 mm
154.9 mm
203 mm
Dimensione della confezione:
Pezzo
Descrizione
- Keep it colorful- this material runs through the BradyJet J4000 or J5000 Inkjet printer for high-resolution, full-color printing
- An ink cartridge is required to print this material
Specifica
Specifica
| Autoclavabile | Not Autoclavable |
| Certificazione/Conformità | BS 5609:1986 Section 2 and section 3, Compliant with Directive 2011/65/EU and 2015/863 |
| Tipo di adesivo | Acrylic, Permanent |
| Rivestimento | None |
| Colore | White |
| Finitura | Opaco |
| Formato | Die-cut |
| Materiale | Polypropylene (B-2585) |
| Printable Sides | Single-side Printable |
| Print Technology | Ink Jet |
| Vedi altri risultati |