missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA120569
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
PA1-20569 detects MAS1 in Human samples.
The structure of the MAS1 product indicates that it belongs to the class of receptors that are coupled to GTP-binding proteins and share a conserved structural motif, which is described as a 7-transmembrane segment following the prediction that these hydrophobic segments form membrane-spanning alpha-helices. The MAS1 protein may be a receptor that, when activated, modulates a critical component in a growth-regulating pathway to bring about oncogenic effects.
Specifica
| MAS1 | |
| Polyclonal | |
| Unconjugated | |
| MAS1 | |
| angiotensin-(1-7) receptor; c-mas; MAS; MAS proto-oncogene; MAS1; Mas-1; MAS1 oncogene; Mas1 proto-oncogene; MAS1 proto-oncogene, G protein-coupled receptor; Mas-related G protein-coupled receptor a; MGC119966; MGRA; Proto-oncogene Mas | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4142 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P04201 | |
| MAS1 | |
| Synthetic 17 amino acid peptide from C-terminus of human MAS1. | |
| 25 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto