Learn More
Greiner Bio-One MASTERBLOCK™ 384 Deep Well Polypropylene Conical Bottom Microplates
Free of detectable DNase, RNase, human DNA.
Marca: Greiner Bio-One 781271
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
- Alphanumeric well coding High chemical resistanceHigh temperature resistanceSealable with adhesive films and heat sealer
Service Plan
No
Specifica
V | |
Without Lid | |
Non Binding | |
Polypropylene | |
20 to 225 μL | |
240 μL | |
6 |
Standard | |
16 x 24 | |
Clear | |
384 | |
Sterile | |
Rounded Square | |
Clear |